ATC517296734 Atlantic Research Chemicals Ltd

2-Butynoyl chloride

  • MDL number MFCD27950345
  • Cas Number : 39753-54-3
  • Molecular Weight : 102.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CC10131
Availability details for
Quantity
  • Availability
  • Add to Cart