B27013117185 BioBlocks, Inc.

(6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-acetic acid

  • Cas Number : 68345-67-5
  • Molecular Weight : 251.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart