B27013117256 BioBlocks, Inc.

4-(3-Methoxy-5-trifluoromethyl-benzyl)-piperidine

  • Cas Number : 794500-99-5
  • Molecular Weight : 273.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart