B27054512215 BioBlocks, Inc.

(R*)-2-((R*)-6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-propan-1-ol

  • Cas Number : 142976-45-2
  • Molecular Weight : 251.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart