B27066469701 BioBlocks, Inc.

trans-2-(4-Methoxy-benzylamino)-cyclohexanol

  • Cas Number : 759402-65-8
  • Molecular Weight : 235.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart