B27066469707 BioBlocks, Inc.

2-(6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-propane-1,3-diol

  • Cas Number : 98661-42-8
  • Molecular Weight : 267.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart