B27066469713 BioBlocks, Inc.

2-Hydroxy-6-methoxy-4-methylquinoline

  • Cas Number : 5342-23-4
  • Molecular Weight : 189.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart