B27068753235 BioBlocks, Inc.

trans-6-Amino-cyclohex-3-enecarboxylic acid

  • Cas Number : 97945-19-2
  • Molecular Weight : 141.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart