B27090268170 BioBlocks, Inc.

[2,2-Dimethyl-1-(2-oxo-ethyl)-propyl]-carbamic acid tert-butyl ester

  • Cas Number : 892874-26-9
  • Molecular Weight : 229.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart