B27100468095 BioBlocks, Inc.

2-(5-thioxo-1H-1,2,4-triazol-4(5H)-yl)acetic acid

  • Cas Number : 61336-27-4
  • Molecular Weight : 159.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart