B27100468096 BioBlocks, Inc.

2-(3-methyl-5-thioxo-1H-1,2,4-triazol-4(5H)-yl)acetic acid

  • Cas Number : 42047-33-6
  • Molecular Weight : 173.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart