B27100468098 BioBlocks, Inc.

(3-Thiophen-2-ylmethyl-5-thioxo-1,5-dihydro-[1,2,4]triazol-4-yl)-acetic acid

  • Cas Number : 892874-27-0
  • Molecular Weight : 255.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart