B27128062846 BioBlocks, Inc.

2,7-Dichloroquinoline-3-carbonitrile

  • Cas Number : 158583-91-6
  • Molecular Weight : 223.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  
Availability details for
Quantity
  • Availability
  • Add to Cart