B27128062948 BioBlocks, Inc.

5,7-Dichloroquinoline-3-carboxylic acid

  • Cas Number : 948293-83-2
  • Molecular Weight : 242.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart