B27128063055 BioBlocks, Inc.

7-Methylquinoline-3-carboxylic acid

  • Cas Number : 948291-17-6
  • Molecular Weight : 187.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart