B27128063172 BioBlocks, Inc.

4-Amino-8-methoxyquinoline

  • Cas Number : 220844-65-5
  • Molecular Weight : 174.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart