B27159734870 BioBlocks, Inc.

1-(1-Amino-ethyl)-naphthalen-2-ol hydrochloride

  • Cas Number : 915781-01-0
  • Molecular Weight : 223.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data HCl
Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart