B27159734872 BioBlocks, Inc.

1-(1-Amino-2-methyl-propyl)naphthalen-2-ol hydrochloride

  • Cas Number : 915781-03-2
  • Molecular Weight : 251.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data HCl
Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart