B27159734882 BioBlocks, Inc.

cis-2-Amino-cyclopentanecarboxylic acid hydrochloride

  • Cas Number : 18414-30-7
  • Molecular Weight : 165.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data HCl
Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart