B27159734936 BioBlocks, Inc.

3-(3-Fluoro-4-methoxy-benzyl)-piperidine hydrochloride

  • Cas Number : 1172742-77-6
  • Molecular Weight : 259.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data HCl
Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart