B27159734943 BioBlocks, Inc.

3-(4-Phenoxy-benzyl)-piperidine hydrochloride

  • Cas Number : 1171985-70-8
  • Molecular Weight : 303.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data HCl
Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart