B27159734964 BioBlocks, Inc.

2-(4-Ethoxy-benzyl)-piperidine hydrochloride

  • Cas Number : 1172745-81-1
  • Molecular Weight : 255.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data HCl
Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart