B27159735039 BioBlocks, Inc.

4-Hydroxy-7-methoxy-2-methylquinoline

  • Cas Number : 103624-90-4
  • Molecular Weight : 189.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart