B27159735115 BioBlocks, Inc.

cis-2-Benzylamino-cyclohexanol hydrochloride

  • Cas Number : 1212095-50-5
  • Molecular Weight : 241.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data HCl
Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart