B27181740129 BioBlocks, Inc.

2-Amino-6-bromo-3-methylquinoline hydrochloride

  • Cas Number : 1171728-14-5
  • Molecular Weight : 273.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data HCl
Availability details for
Quantity
  • Availability
  • Add to Cart