B27181740287 BioBlocks, Inc.

2-(5-Methanesulfonyl-3-phenyl-[1,2,4]triazol-4-yl)-acetic acid

  • Cas Number : 892874-30-5
  • Molecular Weight : 281.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart