B27181740342 BioBlocks, Inc.

2-[[(tert-butoxycarbonyl)amino]methyl]thiazole-4-carboxylic acid

  • Cas Number : 71904-80-8
  • Molecular Weight : 258.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart