B27183269151 BioBlocks, Inc.

4-Bromo-5,8-dichloroquinoline

  • Cas Number : 1070879-38-7
  • Molecular Weight : 276.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart