B27183269165 BioBlocks, Inc.

4-Bromo-6-chloro-2-methylquinoline

  • Cas Number : 1070879-50-3
  • Molecular Weight : 256.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart