B27184894451 BioBlocks, Inc.

(1R*,2S*)-2-(Methylamino)-1,2-diphenylethan-1-ol

  • Cas Number : 20616-52-8
  • Molecular Weight : 227.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart