B27184894453 BioBlocks, Inc.

diexo-3-Amino-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid amide

  • Cas Number : 105786-40-1
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart