B27184894460 BioBlocks, Inc.

diexo-(3-Amino-bicyclo[2.2.1]hept-5-en-2-yl)-methanol

  • Cas Number : 95630-78-7
  • Molecular Weight : 139.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart