B27187420952 BioBlocks, Inc.

4-(Pyrrolidin-1-ylmethyl)aniline hydrochloride

  • Cas Number : 866956-98-1
  • Molecular Weight : 212.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data HCl
Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart