B27234196967 BioBlocks, Inc.

3-[(4-amino-phenylamino)-methyl]- piperidine-1-carboxylic acid tert-butyl ester

  • Cas Number : 1159976-35-8
  • Molecular Weight : 305.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart