B27234196968 BioBlocks, Inc.

2-[(4-amino-phenylamino)-methyl]- piperidine-1-carboxylic acid tert-butyl ester

  • Cas Number : 1159976-36-9
  • Molecular Weight : 305.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart