B27234196973 BioBlocks, Inc.

Ethyl 4-chloro-6-methyl-2-quinazolinecarboxylate

  • Cas Number : 620957-95-1
  • Molecular Weight : 250.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart