B27238958303 BioBlocks, Inc.

5,7-Difluoro-4-hydroxy-2-methylquinoline

  • Cas Number : 288151-40-6
  • Molecular Weight : 195.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart