B27244052801 BioBlocks, Inc.

3-Bromo-4-chloro-6-methylquinoline

  • Cas Number : 1204810-41-2
  • Molecular Weight : 256.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart