B27244052805 BioBlocks, Inc.

3-Bromo-4-chloro-8-methylquinoline

  • Cas Number : 1204810-49-0
  • Molecular Weight : 256.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart