B27244052812 BioBlocks, Inc.

3-Bromo-4-chloro-7-methoxyquinoline

  • Cas Number : 1203579-63-8
  • Molecular Weight : 272.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart