B27333214529 BioBlocks, Inc.

3-Bromo-2,8-dimethyl-4-hydroxyquinoline

  • Cas Number : 909208-88-4
  • Molecular Weight : 252.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart