B27336302530 BioBlocks, Inc.

5-chloro-3-(3-methoxyphenyl)-1,2,4-thiadiazole

  • Cas Number : 946418-99-1
  • Molecular Weight : 226.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart