B27362932982 BioBlocks, Inc.

3-Amino-7-bromoquinoline dihydrochloride

  • Cas Number : 1266226-05-4
  • Molecular Weight : 295.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data HCl
Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart