B27362933020 BioBlocks, Inc.

trans-2-(Methylamino)indan-1-ol

  • Cas Number : 23671-90-1
  • Molecular Weight : 163.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart