B27390499927 BioBlocks, Inc.

cis-tert-butyl 3-(4-nitrophenyl)aziridine-2-carboxylate

  • Cas Number : 1431364-32-7
  • Molecular Weight : 264.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Hungary
Availability details for
Quantity
  • Availability
  • Add to Cart