CH4005788756 ChemBridge Corporation

N-(tert-butyl)-6-chloro-N'-(4-methylphenyl)-1,3,5-triazine-2,4-diamine

  • MDL number MFCD00422444
  • Cas Number : 299929-72-9
  • Molecular Weight : 291.7792

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5101465

Safety Information

MSDS https://www.hit2lead.com/sds/5101465_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart