CH4005789198 ChemBridge Corporation

2-(4-methylphenyl)-1H-indene-1,3(2H)-dione

  • MDL number MFCD00176195
  • Cas Number : 7561-48-0
  • Molecular Weight : 236.2653

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5103385

Safety Information

MSDS https://www.hit2lead.com/sds/5103385_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart