CH4005795319 ChemBridge Corporation

2-(2-hydroxyphenoxy)acetamide

  • MDL number MFCD00547110
  • Cas Number : 34919-87-4
  • Molecular Weight : 167.162

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5144033

Safety Information

MSDS https://www.hit2lead.com/sds/5144033_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart