CH4005796734 ChemBridge Corporation

2-(ethylsulfonyl)-5-iodothiophene

  • MDL number MFCD00228334
  • Cas Number : 412965-13-0
  • Molecular Weight : 302.1531

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5153905

Safety Information

MSDS https://www.hit2lead.com/sds/5153905_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart