CH4005797481 ChemBridge Corporation

4,9-dimethoxy-5-oxo-5H-furo[3,2-g]chromene-7-carboxylic acid

  • MDL number MFCD00410875
  • Cas Number : 25944-28-9
  • Molecular Weight : 290.225

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5160161

Safety Information

MSDS https://www.hit2lead.com/sds/5160161_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart